Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: rnft1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_213228.2 | NP_998393.2 | 6 - 7 | 1109 | 76 | |
GenBank mRNA | BC066378.1 | - | 6 - 7 | 1080 | 76 |
Set Membership: |
Amplicon length between 71 and 85 Mammalian Gene Collection Recommended Assay Danio rerio Probe spans exons |