Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: mpdu1b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001002130.1 | NP_001002130.1 | 6 - 7 | 651 | 58 | |
GenBank mRNA | BC071434.1 | - | 6 - 7 | 651 | 58 |
Set Membership: |
Amplicon length less than or equal to 70 3' Most Mammalian Gene Collection Recommended Assay Danio rerio Probe spans exons |