Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: rras2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001017815.1 | NP_001017815.1 | 4 - 5 | 641 | 62 | |
GenBank mRNA | BC092803.1 | - | 4 - 5 | 641 | 62 | |
BC165269.1 | - | 4 - 5 | 436 | 62 |
Set Membership: |
Amplicon length less than or equal to 70 Mammalian Gene Collection Recommended Assay Danio rerio Inventoried Probe spans exons |