Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: slc27a1b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001077248.1 | NP_001070716.1 | 12 - 13 | 1990 | 89 | |
XM_005160200.4 | XP_005160257.1 | 12 - 13 | 2027 | 89 | ||
GenBank mRNA | BC076535.1 | - | 11 - 12 | 1733 | 89 | |
BC109455.1 | - | 4 - 5 | 500 | 89 | ||
BC124752.1 | - | 12 - 13 | 1990 | 89 | ||
BC164558.1 | - | 11 - 12 | 1825 | 89 |
Set Membership: |
Amplicon length between 86 and 100 3' Most Mammalian Gene Collection Danio rerio Probe spans exons |