Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: sptan1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001098488.1 | NP_001091958.1 | 7 - 8 | 1163 | 77 | |
XM_021468823.1 | XP_021324498.1 | 6 - 7 | 1083 | 77 | ||
XM_021468824.1 | XP_021324499.1 | 7 - 8 | 1215 | 77 | ||
XM_021468825.1 | XP_021324500.1 | 7 - 8 | 1294 | 77 | ||
XM_021468826.1 | XP_021324501.1 | 7 - 8 | 1101 | 77 | ||
XM_021468827.1 | XP_021324502.1 | 6 - 7 | 1083 | 77 | ||
XM_021468828.1 | XP_021324503.1 | 6 - 7 | 1083 | 77 | ||
XM_021468829.1 | XP_021324504.1 | 6 - 7 | 1083 | 77 | ||
XM_021468830.1 | XP_021324505.1 | 6 - 7 | 1083 | 77 | ||
XM_021468831.1 | XP_021324506.1 | 6 - 7 | 1083 | 77 | ||
XM_021468832.1 | XP_021324507.1 | 6 - 7 | 1083 | 77 | ||
XM_021468833.1 | XP_021324508.1 | 6 - 7 | 1083 | 77 | ||
XM_021468834.1 | XP_021324509.1 | 6 - 7 | 1083 | 77 | ||
GenBank mRNA | BC093297.1 | - | 7 - 8 | 1152 | 77 | |
EF375552.1 | - | 7 - 8 | 1163 | 77 |
Set Membership: |
Amplicon length between 71 and 85 Mammalian Gene Collection Detect Genomic DNA Danio rerio Probe spans exons |