Search Thermo Fisher Scientific
- Order Status
- Quick Order
-
Don't have an account ? Create Account
Search Thermo Fisher Scientific
Gene Symbol: kita | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_131053.1 | NP_571128.1 | 13 - 14 | 2019 | 87 | |
XM_009294634.3 | XP_009292909.1 | 13 - 14 | 2064 | 87 | ||
XM_009294635.3 | XP_009292910.1 | 13 - 14 | 2094 | 87 | ||
GenBank mRNA | AF153446.1 | - | 13 - 14 | 2019 | 87 | |
BC162502.1 | - | 13 - 14 | 2034 | 87 | ||
CU458898.1 | - | 13 - 14 | 1970 | 87 |
Set Membership: |
Amplicon length between 86 and 100 Mammalian Gene Collection Detect Genomic DNA Danio rerio Probe spans exons |