Search Thermo Fisher Scientific
- Contáctenos
- Orden Rápida
-
¿No tiene una cuenta? Crear una cuenta
Search Thermo Fisher Scientific
Gene Symbol: prdm1a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_199515.2 | NP_955809.2 | 4 - 5 | 762 | 62 | |
GenBank mRNA | AY497217.1 | - | 4 - 5 | 783 | 62 | |
AY841759.1 | - | 4 - 5 | 791 | 62 | ||
BC080259.1 | - | 4 - 5 | 762 | 62 |
Set Membership: |
Amplicon length less than or equal to 70 Mammalian Gene Collection Recommended Assay Danio rerio Probe spans exons |