Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: rab5aa | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_201485.1 | NP_958893.1 | 5 - 6 | 731 | 77 | |
GenBank mRNA | BC047803.1 | - | 5 - 6 | 731 | 77 | |
BC063966.1 | - | 5 - 6 | 726 | 77 | ||
BC165915.1 | - | 4 - 5 | 571 | 77 |
Set Membership: |
Amplicon length between 71 and 85 3' Most Mammalian Gene Collection Danio rerio Probe spans exons |