Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ilf3b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_212599.1 | NP_997764.1 | 18 - 19 | 2465 | 66 | |
XM_005163739.4 | XP_005163796.1 | 18 - 19 | 2507 | 66 | ||
XM_005163741.3 | XP_005163798.1 | 18 - 19 | 2534 | 66 | ||
XM_009299225.3 | XP_009297500.1 | 17 - 18 | 2445 | 66 | ||
GenBank mRNA | BC067172.1 | - | 18 - 19 | 2465 | 66 |
Set Membership: |
Amplicon length less than or equal to 70 3' Most Mammalian Gene Collection Danio rerio Probe spans exons |