Search Thermo Fisher Scientific
- Contáctenos
- Orden Rápida
-
¿No tiene una cuenta? Crear una cuenta
Search Thermo Fisher Scientific
Gene Symbol: pcdh15b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001012486.1 | NP_001012504.1 | 29 - 30 | 4052 | 94 | |
XM_009306437.3 | XP_009304712.1 | 29 - 30 | 4451 | 94 | ||
XM_009306438.3 | XP_009304713.1 | 28 - 29 | 4445 | 94 | ||
XM_009306439.3 | XP_009304714.1 | 29 - 30 | 4451 | 94 | ||
XM_009306440.3 | XP_009304715.1 | 29 - 30 | 4451 | 94 | ||
GenBank mRNA | AY772391.1 | - | 29 - 30 | 4052 | 94 |
Set Membership: |
Amplicon length between 86 and 100 3' Most Danio rerio Probe spans exons |