Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: gpia | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_144763.1 | NP_658909.1 | 15 - 16 | 1486 | 62 | |
GenBank mRNA | AJ306395.1 | - | 15 - 16 | 1486 | 62 | |
BC044450.1 | - | 15 - 16 | 1472 | 62 | ||
BC083507.1 | - | 15 - 16 | 1470 | 62 | ||
BC165764.1 | - | 15 - 16 | 1427 | 62 |
Set Membership: |
Amplicon length less than or equal to 70 Mammalian Gene Collection Recommended Assay Danio rerio Probe spans exons |