Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: tfdp1b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_200776.2 | NP_957070.2 | 10 - 11 | 1202 | 97 | |
XM_005167988.4 | XP_005168045.1 | 10 - 11 | 1209 | 97 | ||
XM_009305148.3 | XP_009303423.1 | 10 - 11 | 1242 | 97 | ||
GenBank mRNA | BC059610.1 | - | 10 - 11 | 1164 | 97 | |
EH562533.1 | - | 2 - 3 | 480 | 97 |
Set Membership: |
Amplicon length between 86 and 100 3' Most Mammalian Gene Collection Danio rerio Probe spans exons |