Search Thermo Fisher Scientific
- Contact Us
- Quick Order
-
Don't have an account ? Create Account
Search Thermo Fisher Scientific
Gene Symbol: rhot1a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_212704.1 | NP_997869.1 | 11 - 12 | 1169 | 91 | |
XM_009299425.3 | XP_009297700.1 | 11 - 12 | 1198 | 91 | ||
XM_009299426.3 | XP_009297701.1 | 11 - 12 | 1197 | 91 | ||
XM_009299427.3 | XP_009297702.1 | 11 - 12 | 1198 | 91 | ||
GenBank mRNA | BC044431.1 | - | 11 - 12 | 1164 | 91 | |
BC068190.1 | - | 11 - 12 | 1169 | 91 |
Set Membership: |
Amplicon length between 86 and 100 Mammalian Gene Collection Detect Genomic DNA Danio rerio Probe spans exons |