Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: stk25b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_213308.1 | NP_998473.1 | 8 - 9 | 1328 | 80 | |
XM_005171203.4 | XP_005171260.1 | 8 - 9 | 1389 | 80 | ||
GenBank mRNA | BC045867.1 | - | 8 - 9 | 1304 | 80 | |
BC066512.1 | - | 8 - 9 | 1328 | 80 | ||
BC164728.1 | - | 8 - 9 | 1061 | 80 |
Set Membership: |
Amplicon length between 71 and 85 Mammalian Gene Collection Danio rerio Probe spans exons |