Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: pdss2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001002351.1 | NP_001002351.1 | 3 - 4 | 875 | 73 | |
GenBank mRNA | BC075908.1 | - | 3 - 4 | 875 | 73 |
Set Membership: |
Amplicon length between 71 and 85 Mammalian Gene Collection Recommended Assay Danio rerio Probe spans exons |