Search Thermo Fisher Scientific
- Contact Us
- Quick Order
-
Don't have an account ? Create Account
Search Thermo Fisher Scientific
Gene Symbol: selenoh | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_178133.3 | NP_835234.2 | 1 | 500 | 70 | |
GenBank mRNA | AY216587.1 | - | 1 | 346 | 70 | |
CT670471.2 | - | 1 | 328 | 70 | ||
EH510258.1 | - | 1 | 308 | 70 |
Gene Symbol: cenpa | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001164240.1 | NP_001157712.1 | 1 | 1440 | 70 | |
GenBank mRNA | BC044483.1 | - | 1 | 1440 | 70 | |
BC153308.1 | - | 1 | 364 | 70 |
Set Membership: |
Amplicon length less than or equal to 70 3' Most Mammalian Gene Collection Detect Genomic DNA Danio rerio Both primers and probe map within a single exon |