Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: atp5f1d | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_199968.1 | NP_956262.1 | 1 - 2 | 176 | 87 | |
GenBank mRNA | BC049434.1 | - | 1 - 2 | 220 | 87 | |
BC059627.1 | - | 1 - 2 | 176 | 87 | ||
BC065042.1 | - | 1 - 2 | 170 | 87 |
Set Membership: |
Amplicon length between 86 and 100 Mammalian Gene Collection Danio rerio Probe spans exons |