Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: asic1b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_214791.1 | NP_999956.1 | 6 - 7 | 1397 | 77 | |
XM_005161666.4 | XP_005161723.1 | 6 - 7 | 1681 | 77 | ||
XM_005161667.4 | XP_005161724.1 | 7 - 8 | 2133 | 77 | ||
GenBank mRNA | AJ609615.1 | - | 6 - 7 | 1397 | 77 | |
BC162362.1 | - | 6 - 7 | 1366 | 77 | ||
BC162368.1 | - | 6 - 7 | 1366 | 77 |
Set Membership: |
Amplicon length between 71 and 85 Mammalian Gene Collection Danio rerio Probe spans exons |