Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: impact | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001005595.1 | NP_001005595.1 | 11 | 1054 | 133 | |
XM_017353115.2 | XP_017208604.1 | 12 | 1124 | 133 | ||
XM_017353116.1 | XP_017208605.1 | 12 | 1125 | 133 | ||
XM_017353117.2 | XP_017208606.1 | 12 | 1105 | 133 | ||
XM_017353118.1 | XP_017208607.1 | 12 | 1075 | 133 | ||
XM_017353119.2 | XP_017208608.1 | 9 | 1009 | 133 | ||
XM_021469317.1 | XP_021324992.1 | 10 | 963 | 133 | ||
XM_021469318.1 | XP_021324993.1 | 8 | 860 | 133 | ||
GenBank mRNA | BC081507.1 | - | 11 | 1054 | 133 | |
BC152160.1 | - | 12 | 1110 | 133 |
Set Membership: |
Amplicon length greater than or equal to 101 3' Most Mammalian Gene Collection Detect Genomic DNA Recommended Assay Danio rerio Both primers and probe map within a single exon |