Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ass1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001004603.1 | NP_001004603.1 | 3250 | 119 | ||
GenBank mRNA | BC081578.1 | - | 3250 | 119 | ||
BC154282.1 | - | 3253 | 119 |
Set Membership: |
Amplicon length greater than or equal to 101 3' Most Mammalian Gene Collection Recommended Assay Danio rerio |