Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: gfra1a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_131730.1 | NP_571805.1 | 8 | 1346 | 96 | |
GenBank mRNA | AY436320.1 | - | 8 | 1346 | 96 |
Set Membership: |
Amplicon length between 86 and 100 3' Most Detect Genomic DNA Recommended Assay Danio rerio Both primers and probe map within a single exon |