Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: dpy19l1l | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001002868.1 | NP_001002868.1 | 6 - 7 | 792 | 133 | |
GenBank mRNA | AY648728.1 | - | 6 - 7 | 792 | 133 |
Set Membership: |
Amplicon length greater than or equal to 101 Detect Genomic DNA Danio rerio Probe spans exons |