Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: arrdc1a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001004561.1 | NP_001004561.1 | 2 - 3 | 313 | 87 | |
XM_005171783.4 | XP_005171840.1 | 2 - 3 | 298 | 87 | ||
GenBank mRNA | BC081631.1 | - | 2 - 3 | 267 | 87 | |
BC152620.1 | - | 2 - 3 | 282 | 87 |
Set Membership: |
Amplicon length between 86 and 100 Mammalian Gene Collection Detect Genomic DNA Danio rerio Probe spans exons |