Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: osr2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001017694.1 | NP_001017694.1 | 3 - 4 | 824 | 52 | |
GenBank mRNA | BC093148.1 | - | 3 - 4 | 824 | 52 | |
BC165157.1 | - | 2 - 3 | 679 | 52 |
Set Membership: |
Amplicon length less than or equal to 70 3' Most Mammalian Gene Collection Danio rerio Probe spans exons |