Search Thermo Fisher Scientific
- Order Status
- Quick Order
-
Don't have an account ? Create Account
Search Thermo Fisher Scientific
Gene Symbol: aldoca | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001029952.1 | NP_001025123.1 | 6 - 7 | 709 | 67 | |
XM_009291564.3 | XP_009289839.1 | 6 - 7 | 668 | 67 | ||
GenBank mRNA | BC098624.1 | - | 6 - 7 | 709 | 67 |
Set Membership: |
Amplicon length less than or equal to 70 Mammalian Gene Collection Danio rerio Probe spans exons |