Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: cdc37l1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001109826.2 | NP_001103296.2 | 6 - 7 | 994 | 146 | |
GenBank mRNA | BC090726.1 | - | 1 - 2 | 102 | 146 | |
BC152570.1 | - | 6 - 7 | 994 | 146 |
Set Membership: |
Amplicon length greater than or equal to 101 3' Most Mammalian Gene Collection Danio rerio Probe spans exons |