Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ftr24 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001365968.1 | NP_001352897.1 | 6 - 7 | 1265 | 73 | |
GenBank mRNA | BC098890.1 | - | 6 - 7 | 1163 | 73 |
Set Membership: |
Amplicon length between 71 and 85 3' Most Mammalian Gene Collection Recommended Assay Danio rerio Probe spans exons |