Search Thermo Fisher Scientific
- Contáctenos
- Orden Rápida
-
¿No tiene una cuenta? Crear una cuenta
Search Thermo Fisher Scientific
Gene Symbol: arid3b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001083002.1 | NP_001076471.1 | 7 - 8 | 1653 | 104 | |
XM_005163128.4 | XP_005163185.1 | 7 - 8 | 1845 | 104 | ||
XM_009298079.3 | XP_009296354.1 | 7 - 8 | 1636 | 104 | ||
GenBank mRNA | BC131867.1 | - | 7 - 8 | 1653 | 104 |
Set Membership: |
Amplicon length greater than or equal to 101 Mammalian Gene Collection Detect Genomic DNA Danio rerio Probe spans exons |