Search Thermo Fisher Scientific
- Contact Us
- Quick Order
-
Don't have an account ? Create Account
Search Thermo Fisher Scientific
Gene Symbol: bms1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001111150.2 | NP_001104620.2 | 3 - 4 | 519 | 81 | |
GenBank mRNA | BC078263.1 | - | 3 - 4 | 512 | 81 | |
BC134022.1 | - | 3 - 4 | 514 | 81 | ||
BC155256.1 | - | 3 - 4 | 500 | 81 | ||
BC155771.1 | - | 3 - 4 | 497 | 81 |
Set Membership: |
Amplicon length between 71 and 85 Mammalian Gene Collection Danio rerio Probe spans exons |