Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: cadm1a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001113551.2 | NP_001107023.2 | 6 - 7 | 703 | 111 | |
XM_005161197.4 | XP_005161254.1 | 6 - 7 | 758 | 111 | ||
XM_005161198.4 | XP_005161255.1 | 6 - 7 | 757 | 111 | ||
XM_009295329.3 | XP_009293604.1 | 5 - 6 | 740 | 111 | ||
XM_021468894.1 | XP_021324569.1 | 6 - 7 | 758 | 111 | ||
GenBank mRNA | EU182349.2 | - | 6 - 7 | 703 | 111 | |
EU182350.2 | - | 6 - 7 | 703 | 111 | ||
EU182351.2 | - | 6 - 7 | 703 | 111 |
Set Membership: |
Amplicon length greater than or equal to 101 Danio rerio Probe spans exons |