Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: cadm1a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001113551.2 | NP_001107023.2 | 7 - 8 | 801 | 76 | |
XM_005161197.4 | XP_005161254.1 | 7 - 8 | 856 | 76 | ||
XM_005161198.4 | XP_005161255.1 | 7 - 8 | 855 | 76 | ||
XM_009295329.3 | XP_009293604.1 | 6 - 7 | 838 | 76 | ||
XM_021468894.1 | XP_021324569.1 | 7 - 8 | 856 | 76 | ||
GenBank mRNA | EU182349.2 | - | 7 - 8 | 801 | 76 | |
EU182350.2 | - | 7 - 8 | 801 | 76 | ||
EU182351.2 | - | 7 - 8 | 801 | 76 |
Set Membership: |
Amplicon length between 71 and 85 Danio rerio Probe spans exons |