Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ccnd1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_131025.4 | NP_571100.1 | 2 - 3 | 691 | 73 | |
GenBank mRNA | AF365874.1 | - | 2 - 3 | 629 | 73 | |
BC075743.1 | - | 2 - 3 | 648 | 73 | ||
BC152628.1 | - | 2 - 3 | 647 | 73 | ||
CT624760.2 | - | 1 - 2 | 165 | 73 | ||
EH579652.1 | - | 2 - 3 | 681 | 73 | ||
X87581.1 | - | 609 | 73 |
Set Membership: |
Amplicon length between 71 and 85 Mammalian Gene Collection Detect Genomic DNA Danio rerio Probe spans exons |