Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: med29 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_173271.1 | NP_775378.1 | 1 - 2 | 165 | 82 | |
GenBank mRNA | AF506210.1 | - | 1 - 2 | 165 | 82 | |
BC094992.1 | - | 1 - 2 | 161 | 82 | ||
BC164953.1 | - | 1 - 2 | 186 | 82 |
Set Membership: |
Amplicon length between 71 and 85 Mammalian Gene Collection Detect Genomic DNA Danio rerio Probe spans exons |