Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: mmp14b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_194414.1 | NP_919395.1 | 9 - 10 | 1774 | 79 | |
XM_005171310.4 | XP_005171367.1 | 10 - 11 | 1702 | 79 | ||
GenBank mRNA | AY324197.1 | - | 9 - 10 | 1774 | 79 | |
BC162135.1 | - | 9 - 10 | 1666 | 79 |
Set Membership: |
Amplicon length between 71 and 85 3' Most Mammalian Gene Collection Recommended Assay Danio rerio Probe spans exons |