Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: higd1a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_200100.2 | NP_956394.1 | 2 - 3 | 184 | 98 | |
GenBank mRNA | AB094785.1 | - | 2 - 3 | 153 | 98 | |
AB118157.1 | - | 2 - 3 | 122 | 98 | ||
BC062837.1 | - | 2 - 3 | 135 | 98 | ||
EH437255.1 | - | 2 - 3 | 183 | 98 |
Set Membership: |
Amplicon length between 86 and 100 3' Most Mammalian Gene Collection Detect Genomic DNA Recommended Assay Danio rerio Probe spans exons |