Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: arhgap29b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_200111.2 | NP_956405.2 | 16 - 17 | 2142 | 79 | |
XM_005163312.4 | XP_005163369.1 | 16 - 17 | 2250 | 79 | ||
XM_005163314.4 | XP_005163371.1 | 16 - 17 | 2233 | 79 | ||
XM_005163315.4 | XP_005163372.1 | 14 - 15 | 2063 | 79 | ||
GenBank mRNA | BC059184.1 | - | 16 - 17 | 2135 | 79 |
Set Membership: |
Amplicon length between 71 and 85 Mammalian Gene Collection Danio rerio Probe spans exons |