Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: dnajc17 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_200246.2 | NP_956540.2 | 7 - 8 | 658 | 65 | |
GenBank mRNA | BC047828.1 | - | 7 - 8 | 562 | 65 |
Set Membership: |
Amplicon length less than or equal to 70 Mammalian Gene Collection Recommended Assay Danio rerio Probe spans exons |