Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: tiprl | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001328426.1 | NP_001315355.1 | 3 - 4 | 553 | 83 | |
GenBank mRNA | BC057515.1 | - | 3 - 4 | 337 | 83 | |
BC165159.1 | - | 2 - 3 | 329 | 83 |
Set Membership: |
Amplicon length between 71 and 85 Mammalian Gene Collection Danio rerio Probe spans exons |