Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ccdc85a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001320191.1 | NP_001307120.1 | 1 - 2 | 641 | 62 | |
NR_135163.1 | 1 - 2 | 641 | 62 | |||
XM_009307276.3 | XP_009305551.1 | 1 - 2 | 649 | 62 | ||
GenBank mRNA | BC155335.1 | - | 1 - 2 | 613 | 62 | |
EH432792.1 | - | 1 - 2 | 667 | 62 |
Set Membership: |
Amplicon length less than or equal to 70 Mammalian Gene Collection Danio rerio Probe spans exons |