Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: kcmf1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_200952.2 | NP_957246.1 | 6 - 7 | 1040 | 71 | |
GenBank mRNA | AY394941.1 | - | 6 - 7 | 1040 | 71 | |
BC053288.1 | - | 6 - 7 | 1006 | 71 |
Set Membership: |
Amplicon length between 71 and 85 3' Most Mammalian Gene Collection Detect Genomic DNA Danio rerio Probe spans exons |