Search Thermo Fisher Scientific
- Contact Us
- Quick Order
-
Don't have an account ? Create Account
Search Thermo Fisher Scientific
Gene Symbol: hey1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_212561.1 | NP_997726.1 | 3 - 4 | 290 | 142 | |
GenBank mRNA | AJ510221.1 | - | 3 - 4 | 290 | 142 | |
BC095317.1 | - | 3 - 4 | 311 | 142 | ||
BC164111.1 | - | 3 - 4 | 279 | 142 |
Set Membership: |
Amplicon length greater than or equal to 101 Mammalian Gene Collection Detect Genomic DNA Danio rerio Probe spans exons |