Search Thermo Fisher Scientific
- Order Status
- Quick Order
-
Don't have an account ? Create Account
Search Thermo Fisher Scientific
Gene Symbol: rpl17 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001243037.2 | NP_001229966.1 | 3 - 4 | 125 | 71 | |
NM_001319650.1 | NP_001306579.1 | 3 - 4 | 294 | 71 | ||
GenBank mRNA | BC055097.1 | - | 2 - 3 | 86 | 71 | |
CF996097.1 | - | 2 - 3 | 137 | 71 | ||
EB874078.1 | - | 3 - 4 | 116 | 71 | ||
EH488885.1 | - | 3 - 4 | 168 | 71 | ||
EH543457.1 | - | 3 - 4 | 155 | 71 |
Set Membership: |
Amplicon length between 71 and 85 Mammalian Gene Collection Detect Genomic DNA Danio rerio Probe spans exons |