Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TPPP | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_001231863.5 | XP_001231864.1 | 2 - 3 | 405 | 110 | |
XM_004935074.3 | XP_004935131.3 | 3 - 4 | 628 | 110 | ||
XM_015275922.2 | XP_015131408.1 | 2 - 3 | 394 | 110 | ||
XM_025147612.1 | XP_025003380.1 | 2 - 3 | 369 | 110 | ||
XM_418894.6 | XP_418894.3 | 3 - 4 | 510 | 110 | ||
GenBank mRNA | BX932523.2 | - | 3 - 4 | 475 | 110 | |
BX932684.1 | - | 3 - 4 | 495 | 110 | ||
BX934333.2 | - | 3 - 4 | 499 | 110 | ||
BX935488.2 | - | 2 - 3 | 373 | 110 |
Set Membership: |
Amplicon length greater than or equal to 101 Recommended Assay Gallus gallus Probe spans exons |