Search Thermo Fisher Scientific
- Contáctenos
- Orden Rápida
-
¿No tiene una cuenta? Crear una cuenta
Search Thermo Fisher Scientific
Gene Symbol: ZMAT5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001277513.1 | NP_001264442.1 | 2 - 3 | 68 | ||
NM_001277514.1 | NP_001264443.1 | 2 - 3 | 257 | 68 | ||
XM_015294845.2 | XP_015150331.1 | 2 - 3 | 820 | 68 | ||
XM_015294846.2 | XP_015150332.1 | 2 - 3 | 253 | 68 | ||
XM_015294847.2 | XP_015150333.1 | 1 - 2 | 1943 | 68 | ||
XM_025155506.1 | XP_025011274.1 | 1 - 2 | 1944 | 68 | ||
GenBank mRNA | BX930800.2 | - | 2 - 3 | 68 | ||
BX930832.1 | - | 2 - 3 | 68 | |||
BX930880.2 | - | 2 - 3 | 257 | 68 | ||
BX933282.1 | - | 2 - 3 | 259 | 68 | ||
BX933329.1 | - | 1 - 2 | 254 | 68 | ||
BX950293.1 | - | 2 - 3 | 68 |
Set Membership: |
Amplicon length less than or equal to 70 Recommended Assay Gallus gallus Probe spans exons |