Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: FAM173A | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001277151.1 | NP_001264080.1 | 3 - 4 | 415 | 96 | |
NM_001277153.1 | NP_001264082.1 | 2 - 3 | 310 | 96 | ||
NM_001277162.1 | NP_001264091.1 | 3 - 4 | 415 | 96 | ||
XM_015294663.2 | XP_015150149.1 | 3 - 4 | 419 | 96 | ||
GenBank mRNA | BU241996.1 | - | 3 - 4 | 415 | 96 | |
BX930158.1 | - | 3 - 4 | 405 | 96 | ||
BX930472.1 | - | 3 - 4 | 408 | 96 | ||
BX930799.2 | - | 1 - 2 | 126 | 96 | ||
BX931208.2 | - | 3 - 4 | 404 | 96 | ||
BX932017.2 | - | 3 - 4 | 398 | 96 | ||
BX933014.1 | - | 1 - 2 | 126 | 96 | ||
BX935502.2 | - | 3 - 4 | 414 | 96 | ||
BX935727.2 | - | 2 - 3 | 303 | 96 | ||
BX950531.1 | - | 3 - 4 | 411 | 96 |
Set Membership: |
Amplicon length between 86 and 100 3' Most Detect Genomic DNA Gallus gallus Probe spans exons |