Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ERP29 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001276324.1 | NP_001263253.1 | 1 - 2 | 148 | 85 | |
GenBank mRNA | AJ720735.1 | - | 1 - 2 | 149 | 85 | |
BU401484.1 | - | 1 - 2 | 120 | 85 |
Set Membership: |
Amplicon length between 71 and 85 Recommended Assay Gallus gallus Probe spans exons |