Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: BCL10 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001201487.2 | NP_001188416.2 | 2 - 3 | 407 | 120 | |
GenBank mRNA | BU113828.1 | - | 2 - 3 | 339 | 120 | |
BX929269.1 | - | 2 - 3 | 407 | 120 | ||
HQ650869.1 | - | 2 - 3 | 365 | 120 |
Set Membership: |
Amplicon length greater than or equal to 101 3' Most Gallus gallus Probe spans exons |