Search Thermo Fisher Scientific
- Order Status
- Quick Order
-
Don't have an account ? Create Account
Search Thermo Fisher Scientific
Gene Symbol: NTN3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001277605.1 | NP_001264534.1 | 3 - 4 | 1346 | 101 | |
XM_015294747.2 | XP_015150233.1 | 4 - 5 | 1978 | 101 | ||
GenBank mRNA | L34550.1 | - | 3 - 4 | 1288 | 101 |
Set Membership: |
Amplicon length greater than or equal to 101 Gallus gallus Probe spans exons |