Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: LECT1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_204810.1 | NP_990141.1 | 3 - 4 | 360 | 67 | |
GenBank mRNA | AF027380.1 | - | 3 - 4 | 359 | 67 | |
AF138280.1 | - | 3 - 4 | 360 | 67 |
Set Membership: |
Amplicon length less than or equal to 70 Recommended Assay Gallus gallus Probe spans exons |