Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PGA5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_204878.1 | NP_990209.1 | 6 - 7 | 800 | 62 | |
XM_025150584.1 | XP_025006352.1 | 6 - 7 | 848 | 62 | ||
XM_025150585.1 | XP_025006353.1 | 6 - 7 | 848 | 62 | ||
GenBank mRNA | AB025281.1 | - | 6 - 7 | 800 | 62 |
Set Membership: |
Amplicon length less than or equal to 70 Detect Genomic DNA Recommended Assay Gallus gallus Probe spans exons |